2-Chloro-4-(cyclopent-1-en-1-yl)-5-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

124607-1G 124607-5G 124607-25G
77464-372EA 60.77 USD
77464-372 77487-252 77498-072
2-Chloro-4-(cyclopent-1-en-1-yl)-5-fluoropyridine ≥97%
2-Chloro-4-(cyclopent-1-en-1-yl)-5-fluoropyridine

Precaución: For research use only

Formula: C₁₀H₉ClFN
Storage Temperature: Ambient
MDL Number: MFCD34825904
CAS Number: 2918955-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR