1-(2,5-Dichloropyridin-3-yl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

124574-1G 124574-500MG 124574-5G 124574-25G
77464-368EA 415.09 USD
77464-368 77477-158 77487-248 77498-068
1-(2,5-Dichloropyridin-3-yl)cyclopentanol ≥95%
1-(2,5-Dichloropyridin-3-yl)cyclopentanol

Precaución: For research use only

Formula: C₁₀H₁₁Cl₂NO
MW: 232.106 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34825895
CAS Number: 2969413-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR