1-(2-Chloropyridin-3-yl)cyclopentanol ≥97%

Supplier: AOBChem USA

Warning

124522-1G 124522-500MG 124522-5G 124522-25G
77464-364EA 415.09 USD
77464-364 77477-154 77487-244 77498-064
1-(2-Chloropyridin-3-yl)cyclopentanol ≥97%
1-(2-Chloropyridin-3-yl)cyclopentanol

Precaución: For research use only

Formula: C₁₀H₁₂ClNO
MW: 197.661 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34825886
CAS Number: 2969219-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR