1-(6-Bromo-2-fluoropyridin-3-yl)-2-fluoroethanone ≥95%

Supplier: AOBChem USA

Warning

124514-1G 124514-500MG 124514-5G 124514-25G
77464-360EA 415.09 USD
77464-360 77477-148 77487-238 77498-062
1-(6-Bromo-2-fluoropyridin-3-yl)-2-fluoroethanone ≥95%
1-(6-Bromo-2-fluoropyridin-3-yl)-2-fluoroethanone

Precaución: For research use only

Formula: C₇H₄BrF₂NO
Storage Temperature: Ambient
MDL Number: MFCD32851941
CAS Number: 2921687-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR