1-(6-Bromopyridin-3-yl)cyclopentanol ≥97%

Supplier: AOBChem USA

Warning

124129-1G 124129-500MG 124129-5G 124129-25G
77464-356EA 415.09 USD
77464-356 77477-146 77487-236 77498-058
1-(6-Bromopyridin-3-yl)cyclopentanol ≥97%
1-(6-Bromopyridin-3-yl)cyclopentanol

Precaución: For research use only

Formula: C₁₀H₁₂BrNO
Storage Temperature: Ambient
MDL Number: MFCD27930471
CAS Number: 865204-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR