2-(6-Bromo-2-fluoro-3-methoxyphenyl)-1,1,1-trifluoropropan-2-ol ≥95%

Supplier: AOBChem USA

Warning

124101-1G 124101-500MG 124101-5G 124101-25G
77464-352EA 415.09 USD
77464-352 77477-142 77487-232 77498-054
2-(6-Bromo-2-fluoro-3-methoxyphenyl)-1,1,1-trifluoropropan-2-ol ≥95%
2-(6-Bromo-2-fluoro-3-methoxyphenyl)-1,1,1-trifluoropropan-2-ol

Precaución: For research use only

Formula: C₁₀H₉BrF₄O₂
MW: 317.0748 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34784708
CAS Number: 2918832-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR