Methyl 2-(benzyloxy)-5-bromobenzoate ≥96%

Supplier: AOBChem USA

Warning

123976-1G 123976-500MG 123976-5G 123976-25G
77464-348EA 264.71 USD
77464-348 77477-138 77487-228 77498-052
Methyl 2-(benzyloxy)-5-bromobenzoate ≥96%
Methyl 2-(benzyloxy)-5-bromobenzoate

Precaución: For research use only

Formula: C₁₅H₁₃BrO₃
MW: 321.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11112970
CAS Number: 860000-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR