3-Cyano-5-(trifluoromethoxy)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

123522-1G 123522-500MG 123522-5G 123522-25G
77464-344EA 80.34 USD
77464-344 77477-134 77487-222 77498-046
3-Cyano-5-(trifluoromethoxy)benzoic acid ≥95%
3-Cyano-5-(trifluoromethoxy)benzoic acid

Precaución: For research use only

Formula: C₉H₄F₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD28782102
CAS Number: 453565-91-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR