5-Bromo-1-(diethoxymethyl)-2-fluoro-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

123388-1G 123388-500MG 123388-5G 123388-25G
77464-340EA 184.37 USD
77464-340 77477-130 77487-218 77498-044
5-Bromo-1-(diethoxymethyl)-2-fluoro-3-(trifluoromethyl)benzene ≥97%
5-Bromo-1-(diethoxymethyl)-2-fluoro-3-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₁₂H₁₃BrF₄O₂
MW: 345.128 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD34825370
CAS Number: 2918963-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR