3-Ethoxy-2-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

122940-1G 122940-500MG 122940-5G 122940-25G
77464-336EA 252.35 USD
77464-336 77477-126 77487-214 77498-032
3-Ethoxy-2-methylbenzoic acid ≥95%
3-Ethoxy-2-methylbenzoic acid

Precaución: For research use only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02259458
CAS Number: 103323-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR