2-(2-Bromo-3-fluoro-4-methoxyphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

122596-1G 122596-5G 122596-25G
77464-332EA 60.77 USD
77464-332 77487-210 77498-028
2-(2-Bromo-3-fluoro-4-methoxyphenyl)-1,3-dioxolane ≥97%
2-(2-Bromo-3-fluoro-4-methoxyphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₀BrFO₃
MW: 277.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34825003
CAS Number: 2918954-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR