1-(4-Fluoro-3-methoxyphenyl)propan-1-ol ≥97%

Supplier: AOBChem USA

Warning

122032-1G 122032-500MG 122032-5G 122032-25G
77464-328EA 397.58 USD
77464-328 77477-120 77487-206 77498-018
1-(4-Fluoro-3-methoxyphenyl)propan-1-ol ≥97%
1-(4-Fluoro-3-methoxyphenyl)propan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₃FO₂
MW: 184.207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15478771
CAS Number: 82846-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR