1-(2-Fluoro-4-(methylthio)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

121817-1G 121817-500MG 121817-5G
77464-324EA 685.98 USD
77464-324 77477-116 77487-202
1-(2-Fluoro-4-(methylthio)phenyl)ethanone ≥97%
1-(2-Fluoro-4-(methylthio)phenyl)ethanone

Precaución: For research use only

Formula: C₉H₉FOS
MW: 184.231 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28521722
CAS Number: 2968-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR