1-Fluoro-2-iodo-5-methoxy-3-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

120914-1G 120914-500MG 120914-5G 120914-25G
77464-320EA 415.09 USD
77464-320 77477-112 77487-198 77498-008
1-Fluoro-2-iodo-5-methoxy-3-(methoxymethoxy)benzene ≥97%
1-Fluoro-2-iodo-5-methoxy-3-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₉H₁₀FIO₃
MW: 312.077 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34784676
CAS Number: 2901084-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR