(2-(Methoxymethoxy)-3-(Methylthio)phenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

120911-1G 120911-500MG 120911-5G 120911-25G
77464-316EA 415.09 USD
77464-316 77477-108 77487-194 77498-004
(2-(Methoxymethoxy)-3-(Methylthio)phenyl)trimethylsilane ≥97%
(2-(Methoxymethoxy)-3-(Methylthio)phenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₂H₂₀O₂SSi
Storage Temperature: Ambient
MDL Number: MFCD34824252
CAS Number: 2918964-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR