3,5-Dibromo-4-fluorobenzaldehyde ≥96%

Supplier: AOBChem USA

Warning

120650-1G 120650-500MG 120650-5G
77464-312EA 893.01 USD
77464-312 77477-104 77487-188
3,5-Dibromo-4-fluorobenzaldehyde ≥96%
3,5-Dibromo-4-fluorobenzaldehyde

Precaución: For research use only

Formula: C₇H₃Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD28784593
CAS Number: 477535-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR