1-(3-Bromophenyl)ethanone oxime ≥97%

Supplier: AOBChem USA

Warning

120616-1G 120616-500MG 120616-5G 120616-25G
77464-308EA 197.76 USD
77464-308 77477-100 77487-184 77497-998
1-(3-Bromophenyl)ethanone oxime ≥97%
1-(3-Bromophenyl)ethanone oxime

Precaución: For research use only

Formula: C₈H₈BrNO
MW: 214.059 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00219816
CAS Number: 187672-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR