1-(4-(Methylthio)phenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

120603-1G 120603-500MG 120603-5G
77464-300EA 648.9 USD
77464-300 77477-092 77487-176
1-(4-(Methylthio)phenyl)propan-1-ol ≥95%
1-(4-(Methylthio)phenyl)propan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD11617676
CAS Number: 89434-30-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR