(3-Methoxy-4-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

120579-1G 120579-500MG 120579-5G 120579-25G
77464-296EA 415.09 USD
77464-296 77477-088 77487-172 77497-992
(3-Methoxy-4-methylphenyl)(methyl)sulfane ≥95%
(3-Methoxy-4-methylphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₉H₁₂OS
Storage Temperature: Ambient
MDL Number: MFCD22489317
CAS Number: 876497-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR