1-(4-Ethoxy-2,3-difluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

120382-1G 120382-500MG 120382-5G 120382-25G
77464-292EA 415.09 USD
77464-292 77477-084 77487-168 77497-990
1-(4-Ethoxy-2,3-difluorophenyl)-2,2-difluoroethanone ≥95%
1-(4-Ethoxy-2,3-difluorophenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₈F₄O₂
MW: 236.163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34824056
CAS Number: 2969455-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR