2-(3-Iodothiophen-2-yl)-2-methyl-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

119857-1G 119857-500MG 119857-5G 119857-25G
77464-284EA 109.18 USD
77464-284 77477-076 77487-160 77497-982
2-(3-Iodothiophen-2-yl)-2-methyl-1,3-dioxolane ≥97%
2-(3-Iodothiophen-2-yl)-2-methyl-1,3-dioxolane

Precaución: For research use only

Formula: C₈H₉IO₂S
MW: 296.125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34823901
CAS Number: 32969-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR