5-acetyl-4-bromothiophene-2-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

119855-1G 119855-500MG 119855-5G 119855-25G
77464-280EA 156.56 USD
77464-280 77477-072 77487-156 77497-978
5-acetyl-4-bromothiophene-2-carbaldehyde ≥97%
5-acetyl-4-bromothiophene-2-carbaldehyde

Precaución: For research use only

Formula: C₇H₅BrO₂S
Storage Temperature: Ambient
MDL Number: MFCD34823900
CAS Number: 2918967-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR