5-Bromo-8-fluoroquinoline-7-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

119834-1G 119834-500MG 119834-5G 119834-25G
77464-276EA 415.09 USD
77464-276 77477-068 77487-152 77497-974
5-Bromo-8-fluoroquinoline-7-carbaldehyde ≥97%
5-Bromo-8-fluoroquinoline-7-carbaldehyde

Precaución: For research use only

Formula: C₁₀H₅BrFNO
MW: 254.055 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34784520
CAS Number: 2901097-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR