4-(methylthio)isoquinoline ≥97%

Supplier: AOBChem USA

Warning

119651-1G 119651-500MG 119651-5G
77464-268EA 479.98 USD
77464-268 77477-060 77487-144
4-(methylthio)isoquinoline ≥97%
4-(methylthio)isoquinoline

Precaución: For research use only

Formula: C₁₀H₉NS
Storage Temperature: Ambient
MDL Number: MFCD05981989
CAS Number: 38896-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR