1-(3-Chloro-2-fluoro-4-methoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

119598-1G 119598-500MG 119598-5G
77464-264EA 676.71 USD
77464-264 77477-056 77487-140
1-(3-Chloro-2-fluoro-4-methoxyphenyl)ethanone ≥95%
1-(3-Chloro-2-fluoro-4-methoxyphenyl)ethanone

Precaución: For research use only

Formula: C₉H₈ClFO₂
MW: 202.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28640335
CAS Number: 1554889-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR