(3-Isopropyl-4-methoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

119417-1G 119417-500MG 119417-5G 119417-25G
77464-260EA 415.09 USD
77464-260 77477-052 77487-136 77497-966
(3-Isopropyl-4-methoxyphenyl)(methyl)sulfane ≥95%
(3-Isopropyl-4-methoxyphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₁H₁₆OS
MW: 196.309 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34784403
CAS Number: 2918847-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR