Methyl 2-chloro-6-methoxy-3-Methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

119350-1G 119350-500MG 119350-5G
77464-256EA 485.13 USD
77464-256 77477-048 77487-132
Methyl 2-chloro-6-methoxy-3-Methylbenzoate ≥97%
Methyl 2-chloro-6-methoxy-3-Methylbenzoate

Precaución: For research use only

Formula: C₁₀H₁₁ClO₃
MW: 214.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34823778
CAS Number: 2114295-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR