2-Chloro-4-(methoxymethoxy)-1-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

119333-1G 119333-500MG 119333-5G 119333-25G
77464-252EA 82.4 USD
77464-252 77477-044 77487-128 77497-962
2-Chloro-4-(methoxymethoxy)-1-methylbenzene ≥95%
2-Chloro-4-(methoxymethoxy)-1-methylbenzene

Precaución: For research use only

Formula: C₉H₁₁ClO₂
MW: 186.635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27932308
CAS Number: 911718-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR