Butyl(4-fluorophenyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

119260-1G 119260-500MG 119260-5G 119260-25G
77464-248EA 397.58 USD
77464-248 77477-040 77487-124 77497-960
Butyl(4-fluorophenyl)sulfane ≥97%
Butyl(4-fluorophenyl)sulfane

Precaución: For research use only

Formula: C₁₀H₁₃FS
MW: 184.274 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22054907
CAS Number: 1649-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR