2-Bromo-5-fluoro-4-(methylthio)pyridine ≥95%

Supplier: AOBChem USA

Warning

119166-1G 119166-500MG 119166-5G 119166-25G
77464-244EA 415.09 USD
77464-244 77477-036 77487-120 77497-956
2-Bromo-5-fluoro-4-(methylthio)pyridine ≥95%
2-Bromo-5-fluoro-4-(methylthio)pyridine

Precaución: For research use only

Formula: C₆H₅BrFNS
MW: 222.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34823723
CAS Number: 2921684-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR