6-Bromo-4-chlorobenzo[d][1,3]dioxole ≥97%

Supplier: AOBChem USA

Warning

119080-1G 119080-500MG 119080-5G
77464-240EA 506.76 USD
77464-240 77477-032 77487-116
6-Bromo-4-chlorobenzo[d][1,3]dioxole ≥97%
6-Bromo-4-chlorobenzo[d][1,3]dioxole

Precaución: For research use only

Formula: C₇H₄BrClO₂
MW: 235.462 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23709710
CAS Number: 72736-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR