5-Acetylfuran-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

118830-1G 118830-500MG 118830-5G
77464-236EA 580.92 USD
77464-236 77477-028 77487-110
5-Acetylfuran-2-carboxylic acid ≥97%
5-Acetylfuran-2-carboxylic acid

Precaución: For research use only

Formula: C₇H₆O₄
MW: 154.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18431708
CAS Number: 13341-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR