(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118562-1G 118562-500MG 118562-5G 118562-25G
77464-232EA 415.09 USD
77464-232 77477-024 77487-106 77497-948
(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)(phenyl)methanol ≥95%
(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01070798
CAS Number: 351342-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR