Naphthalen-1-yl(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118557-1G 118557-500MG 118557-5G
77464-228EA 471.74 USD
77464-228 77477-020 77487-102
Naphthalen-1-yl(phenyl)methanol ≥95%
Naphthalen-1-yl(phenyl)methanol

Precaución: For research use only

Formula: C₁₇H₁₄O
MW: 234.292 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00029360
CAS Number: 642-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR