(3-Fluoro-5-(trifluoromethyl)phenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118537-1G 118537-500MG 118537-5G 118537-25G
77464-224EA 415.09 USD
77464-224 77477-016 77487-098 77497-942
(3-Fluoro-5-(trifluoromethyl)phenyl)(phenyl)methanol ≥95%
(3-Fluoro-5-(trifluoromethyl)phenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₀F₄O
Storage Temperature: Ambient
MDL Number: MFCD23639435
CAS Number: 1550146-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR