(5-Fluoro-2-methoxyphenyl)(phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

118488-1G 118488-500MG 118488-5G 118488-25G
77464-220EA 397.58 USD
77464-220 77477-012 77487-094 77497-938
(5-Fluoro-2-methoxyphenyl)(phenyl)methanol ≥97%
(5-Fluoro-2-methoxyphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₃FO₂
MW: 232.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16358567
CAS Number: 1275527-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR