(2,4-Dimethylphenyl)(phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

118479-1G 118479-500MG 118479-5G 118479-25G
77464-216EA 350.2 USD
77464-216 77477-008 77487-090 77497-934
(2,4-Dimethylphenyl)(phenyl)methanol ≥97%
(2,4-Dimethylphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₅H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD00962322
CAS Number: 67135-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR