phenyl(4-propylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118470-1G 118470-500MG 118470-5G 118470-25G
77464-212EA 315.18 USD
77464-212 77477-004 77487-086 77497-930
phenyl(4-propylphenyl)methanol ≥95%
phenyl(4-propylphenyl)methanol

Precaución: For research use only

Formula: C₁₆H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD11100904
CAS Number: 51166-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR