4-(Dimethylamino)benzhydrol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (4-(dimethylamino)phenyl)(phenyl)methanol

118468-1G 118468-500MG 118468-5G 118468-25G
77464-208EA 179.22 USD
77464-208 77477-000 77487-082 77497-926
4-(Dimethylamino)benzhydrol ≥95%
4-(Dimethylamino)benzhydrol

Precaución: For research use only

Formula: C₁₅H₁₇NO
Storage Temperature: Ambient
MDL Number: MFCD00963078
CAS Number: 7494-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR