(4-Isopropoxyphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118461-1G 118461-500MG 118461-5G 118461-25G
77464-204EA 415.09 USD
77464-204 77476-996 77487-078 77497-922
(4-Isopropoxyphenyl)(phenyl)methanol ≥95%
(4-Isopropoxyphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₆H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD16358515
CAS Number: 93014-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR