4-tert-Butylbenzhydrol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (4-(tert-Butyl)phenyl)(phenyl)methanol

118459-1G 118459-500MG 118459-5G 118459-25G
77464-200EA 80.34 USD
77464-200 77476-992 77487-074 77497-918
4-tert-Butylbenzhydrol ≥95%
4-tert-Butylbenzhydrol

Precaución: For research use only

Formula: C₁₇H₂₀O
MW: 240.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201248
CAS Number: 22543-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR