(3-Isopropoxyphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118445-1G 118445-500MG 118445-5G 118445-25G
77464-196EA 251.32 USD
77464-196 77476-988 77487-070 77497-912
(3-Isopropoxyphenyl)(phenyl)methanol ≥95%
(3-Isopropoxyphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₆H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD11617519
CAS Number: 1410128-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR