(3-Isopropylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118441-1G 118441-500MG 118441-5G 118441-25G
77464-192EA 415.09 USD
77464-192 77476-984 77487-066 77497-908
(3-Isopropylphenyl)(phenyl)methanol ≥95%
(3-Isopropylphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₆H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD34784103
CAS Number: 108774-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR