3-Methoxybenzhydrol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Methoxyphenyl)(phenyl)methanol

118438-1G 118438-500MG 118438-5G 118438-25G
77464-188EA 120.51 USD
77464-188 77476-980 77487-062 77497-904
3-Methoxybenzhydrol ≥97%
3-Methoxybenzhydrol

Precaución: For research use only

Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201261
CAS Number: 13391-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR