(2-(Methylthio)phenyl)(phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

118435-1G 118435-500MG 118435-5G 118435-25G
77464-184EA 226.6 USD
77464-184 77476-976 77487-058 77497-900
(2-(Methylthio)phenyl)(phenyl)methanol ≥97%
(2-(Methylthio)phenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD21826823
CAS Number: 24845-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR