(2-Ethylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118427-1G 118427-500MG 118427-5G 118427-25G
77464-180EA 415.09 USD
77464-180 77476-972 77487-054 77497-896
(2-Ethylphenyl)(phenyl)methanol ≥95%
(2-Ethylphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₅H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD00598357
CAS Number: 2029-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR