(2-Isopropoxy-4-(trifluoromethyl)phenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

118352-1G 118352-500MG 118352-5G 118352-25G
77464-172EA 415.09 USD
77464-172 77476-964 77487-046 77497-890
(2-Isopropoxy-4-(trifluoromethyl)phenyl)(methyl)sulfane ≥95%
(2-Isopropoxy-4-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₁H₁₃F₃OS
MW: 250.281 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34784079
CAS Number: 2918843-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR