1-(2-Bromo-5-chloropyridin-4-yl)-2-methylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

118313-1G 118313-500MG 118313-5G 118313-25G
77464-168EA 415.09 USD
77464-168 77476-960 77487-042 77497-886
1-(2-Bromo-5-chloropyridin-4-yl)-2-methylpropan-1-ol ≥97%
1-(2-Bromo-5-chloropyridin-4-yl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₉H₁₁BrClNO
MW: 264.547 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34784057
CAS Number: 2901109-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR