1-(5-Chloro-2-fluoro-3-methylphenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

118264-1G 118264-500MG 118264-5G
77464-156EA 778.68 USD
77464-156 77476-948 77487-028
1-(5-Chloro-2-fluoro-3-methylphenyl)-2-methylpropan-1-ol ≥95%
1-(5-Chloro-2-fluoro-3-methylphenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₄ClFO
Storage Temperature: Ambient
MDL Number: MFCD34784013
CAS Number: 2918931-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR