(2-Fluoro-5-iodophenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118221-1G 118221-500MG 118221-5G 118221-25G
77464-152EA 415.09 USD
77464-152 77476-944 77487-024 77497-872
(2-Fluoro-5-iodophenyl)(phenyl)methanol ≥95%
(2-Fluoro-5-iodophenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₃H₁₀FIO
MW: 328.121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34783972
CAS Number: 2918846-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR